2-Bromo-6-chloro-4-fluoroaniline ≥97%

569283-5G 569283-1G 569283-U
220079-670EA 79.91 USD
220079-670 220079-669 220079-671
2-Bromo-6-chloro-4-fluoroaniline ≥97%
2-Bromo-6-chloro-4-fluoroaniline
Formula: C₆H₄BrClFN
MW: 224.46 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 55…56 °C
MDL Number: MFCD00051707
CAS Number: 201849-14-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR