5-Bromo-1-methylimidazole

Synonyms: 5-Bromo-1-methyl-1H-imidazole

568279-25G 568279-5G
220079-180EA 1152.31 USD
220079-180 220079-181
5-Bromo-1-methylimidazole
5-Bromo-1-methylimidazole
Formula: C₄H₅BrN₂
MW: 161 g/mol
MDL Number: MFCD01632218
CAS Number: 1003-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR