3-(Difluoromethoxy)aniline ≥97%

Warning

566446-5G 566446-1G
220077-589EA 183.08 USD
220077-589 220077-588
3-(Difluoromethoxy)aniline ≥97%
3-(Difluoromethoxy)aniline
Formula: C₇H₇F₂NO
MW: 159.14 g/mol
MDL Number: MFCD00236221
CAS Number: 22236-08-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR