4-Bromo-m-toluonitrile ≥97%

566365-25G 566365-5G
220077-583EA 336 USD
220077-583 220077-584
4-Bromo-m-toluonitrile ≥97%
4-Bromo-m-toluonitrile
Formula: C₈H₆BrN
MW: 196.05 g/mol
Boiling Pt: 78…82 °C (0.3 mmHg)
Melting Pt: 55 °C
MDL Number: MFCD00031538
CAS Number: 41963-20-6
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR