m-Toluenesulfonyl chloride

Danger

Synonyms: 3-Toluenesulphonyl chloride, Toluene-3-sulphonyl chloride, Toluene-m-sulphonyl chloride

566357-5G 566357-25G
220077-582EA 87.12 USD
220077-582 220077-581
m-Toluenesulfonyl chloride
m-Toluenesulphonyl chloride
Formula: C₇H₇ClO₂S
MW: 190.65 g/mol
Boiling Pt: 77 - 78 °C (0.3 -rr)
Melting Pt: 11.7 °C
Density: 1.314 g/cm³ (20 °C)
MDL Number: MFCD00051747
CAS Number: 1899-93-0
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR