2-Chloro-6-fluorophenylboronic acid

Synonyms: (2-Chloro-6-fluorophenyl)boronic acid, (2-Chloro-6-fluorophenyl)metaboric acid, 2-Chloro-6-fluorobenzeneboronic acid, Dihydroxy(2-chloro-6-fluorophenyl)borane

566071-5G 566071-1G
220079-091EA 122.31 USD
220079-091 220079-090
2-Chloro-6-fluorophenylboronic acid
2-Chloro-6-fluorophenylboronic acid
Formula: C₆H₅BClFO₂
MW: 174.37 g/mol
MDL Number: MFCD04039892
CAS Number: 313545-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR