4-(Trifluoromethyl)benzenesulfonyl chloride ≥97%

Synonyms: 4-(Trifluoromethyl)benzene-1-sulphonyl chloride

565849-5G 565849-1G
220079-071EA 114.15 USD
220079-071 220079-070
4-(Trifluoromethyl)benzenesulfonyl chloride ≥97%
4-(Trifluoromethyl)benzenesulphonyl chloride
Formula: C₇H₄ClF₃O₂S
MW: 244.62 g/mol
Boiling Pt: 49.5 - 50.5 °C (0.2 -rr)
Melting Pt: 31 - 33 °C
MDL Number: MFCD00042422
CAS Number: 2991-42-6
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR