Anthranilohydrazide ≥97%

Synonyms: 2-Aminobenzoic hydrazide, 2-Aminobenzoic acid hydrazide, 2-Aminobenzohydrazide, 2-Aminobenzoylhydrazide, Anthraniloyl Hydrazine, 2-Aminobenzene-1-carbohydrazide, 2-Aminobenzhydrazide

565490-50G 565490-10G
220079-042EA 196.15 USD
220079-042 220079-041
Anthranilohydrazide ≥97%
Anthranilohydrazide
Formula: C₇H₉N₃O
MW: 151.17 g/mol
Boiling Pt: 378 °C (1013 hPa)
Melting Pt: 123 °C
MDL Number: MFCD00017056
CAS Number: 1904-58-1
EINECS: 217-604-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR