3-Chloro-4-methoxybenzaldehyde ≥97%

Synonyms: 3-Chloro-4-methoxybenzaldehyde

565040-1G 565040-5G
220078-970EA 78.08 USD
220078-970 220078-971
3-Chloro-4-methoxybenzaldehyde ≥97%
3-Chloro-4-methoxybenzaldehyde
Formula: C₈H₇ClO₂
MW: 170.6 g/mol
MDL Number: MFCD00016601
CAS Number: 4903-09-7
EINECS: 225-532-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR