1-Ethyl-3-methylimidazolium hydrogen sulfate, Sigma-Aldrich®

Synonyms: 1-Ethyl-3-methylimidazolium Hydrogen sulphate

56486-100G-F 56486-1KG-F
220077-403EA 263 USD
220077-403 220077-404
1-Ethyl-3-methylimidazolium hydrogen sulfate, Sigma-Aldrich®
1-Ethyl-3-methylimidazolium hydrogen sulphate
Formula: C₆H₁₂N₂O₄S
MW: 208.24 g/mol
MDL Number: MFCD06798195
CAS Number: 412009-61-1
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR