Potassium phenyltrifluoroborate ≥95%

Synonyms: Potassium trifluoro(phenyl)borate, Phenyltrifluoroboric acid potassium salt

563951-5G 563951-1G
220078-243EA 283.85 USD
220078-243 220078-169
Potassium phenyltrifluoroborate ≥95%
Potassium phenyltrifluoroborate
Formula: C₆H₅BF₃K
MW: 184.01 g/mol
MDL Number: MFCD01318172
CAS Number: 153766-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR