2-Bromo-5-fluorobenzoic acid ≥96%

Warning

563498-1G 563498-5G
220078-882EA 464.62 USD
220078-882 220078-883
2-Bromo-5-fluorobenzoic acid ≥96%
2-Bromo-5-fluorobenzoic acid
Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
MDL Number: MFCD00142874
CAS Number: 394-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR