6-Bromo-o-toluidine ≥95%

563048-5G 563048-1G
220078-831EA 252.69 USD
220078-831 220078-830
6-Bromo-o-toluidine ≥95%
6-Bromo-o-toluidine
Formula: C₇H₈BrN
MW: 186.05 g/mol
MDL Number: MFCD04039885
CAS Number: 53848-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR