4-Propylbenzaldehyde ≥97%

Warning

562882-1G 562882-5G
220077-488EA 298.31 USD
220077-488 220078-815
4-Propylbenzaldehyde ≥97%
4-Propylbenzaldehyde
Formula: C₁₀H₁₂O
MW: 148.2 g/mol
Boiling Pt: 240 °C (1013 hPa)
Melting Pt: 19 °C
MDL Number: MFCD00041870
CAS Number: 28785-06-0
EINECS: 249-221-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR