Piperidin-3-ol

Warning

Synonyms: 3-Piperidinol, 3-Hydroxypiperidine

56210-50G 56210-10G
220077-389EA 199.23 USD
220077-389 220077-388
Piperidin-3-ol
Piperidin-3-ol
Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 113…116 °C (1013 hPa)
Melting Pt: 56…59 °C
MDL Number: MFCD00014591
CAS Number: 6859-99-0
EINECS: 229-957-4
UN: 3263
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR