2-Iodo-m-toluic acid ≥97%

560499-1G 560499-5G
220078-637EA 76.03 USD
220078-637 220078-638
2-Iodo-m-toluic acid ≥97%
2-Iodo-m-toluic acid
Formula: C₈H₇IO₂
MW: 262.05 g/mol
MDL Number: MFCD00079764
CAS Number: 108078-14-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR