3-(4-Aminophenyl)propionic acid ≥97%

Synonyms: 4-Aminohydrocinnamic acid, 3-(4-Aminophenyl)propanoic acid

560251-1G 560251-5G
220077-912EA 32.31 USD
220077-912 220077-913
3-(4-Aminophenyl)propionic acid ≥97%
3-(4-Aminophenyl)propionic acid
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD00017118
CAS Number: 2393-17-1
EINECS: 219-245-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR