4-Benzylaniline ≥97%

Synonyms: 4-Aminodiphenylmethane

560235-1G 560235-5G
220078-555EA 25.85 USD
220078-555 220078-556
4-Benzylaniline ≥97%
4-Benzylaniline
Formula: C₁₃H₁₃N
MW: 183.25 g/mol
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 35 - 36 °C
Density: 1.07 g/cm³ (20 °C)
MDL Number: MFCD00047861
CAS Number: 1135-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR