α-Chloro-2,6-difluorotoulene ≥97%

Danger

Synonyms: 2-(Chloromethyl)-1,3-difluorobenzene, 2,6-Difluorobenzyl chloride, 1-(Chloromethyl)-2,6-difluorobenzene

559954-25G 559954-5G
220078-516EA 166.92 USD
220078-516 220078-517
α-Chloro-2,6-difluorotoulene ≥97%
α-Chloro-2,6-difluorotoulene
Formula: C₇H₅ClF₂
MW: 162.57 g/mol
Boiling Pt: 172…173 °C (640 mmHg)
Melting Pt: 32 °C
MDL Number: MFCD00191346
CAS Number: 697-73-4
UN: 1759
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR