3,4,5-Trifluoroaniline ≥98%

Warning

559490-1G 559490-5G
220078-488EA 55.38 USD
220078-488 220078-489
3,4,5-Trifluoroaniline ≥98%
3,4,5-Trifluoroaniline
Formula: C₆H₄F₃N
MW: 147.1 g/mol
MDL Number: MFCD00083522
CAS Number: 163733-96-8
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR