1-(2-Bromoethyl)naphthalene

559318-1G 559318-5G
220078-476EA 105.32 USD
220078-476 220078-477
1-(2-Bromoethyl)naphthalene
1-(2-Bromoethyl)naphthalene
Formula: C₁₂H₁₁Br
MW: 235.12 g/mol
MDL Number: MFCD00037737
CAS Number: 13686-49-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR