α-Bromo-4-chlorotoulene ≥97%

Danger

Synonyms: alpha-Bromo-4-chlorotoluene, 1-(Bromomethyl)-4-chlorobenzene, 4-Chlorobenzyl bromide

558958-100G 558958-25G
220078-442EA 248.77 USD
220078-442 220078-443
α-Bromo-4-chlorotoulene ≥97%
α-Bromo-4-chlorotoulene
Formula: C₇H₆BrCl
MW: 205.48 g/mol
Boiling Pt: 118 - 123 °C (19 -rr)
Melting Pt: 48 - 50 °C
MDL Number: MFCD00040714
CAS Number: 622-95-7
EINECS: 210-760-7
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR