4-Bromo-2-(trifluoromethyl)benzenesulfonyl chloride ≥97%

Synonyms: 4-Bromo-2-(trifluoromethyl)benzene-1-sulphonyl chloride

558664-5G 558664-1G
220078-423EA 152.31 USD
220078-423 220078-422
4-Bromo-2-(trifluoromethyl)benzenesulfonyl chloride ≥97%
4-Bromo-2-(trifluoromethyl)benzenesulphonyl chloride
Formula: C₇H₃BrClF₃O₂S
MW: 323.52 g/mol
Melting Pt: 54 - 58 °C
MDL Number: MFCD03094392
CAS Number: 176225-10-8
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR