5-Fluoro-2-methylbenzenesulfonyl chloride ≥97%

Synonyms: 5-Fluoro-2-methylbenzene-1-sulphonyl chloride, 3-Fluor-6-methylbenzenesulphonyl chloride

558583-1G 558583-5G
220078-411EA 64.83 USD
220078-411 220078-412
5-Fluoro-2-methylbenzenesulfonyl chloride ≥97%
5-Fluoro-2-methylbenzenesulphonyl chloride
Formula: C₇H₆ClFO₂S
MW: 208.64 g/mol
Boiling Pt: 155…158 °C (50 mmHg)
MDL Number: MFCD00052958
CAS Number: 445-05-6
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR