4-Fluoro-2-methylbenzenesulfonyl chloride ≥97%

Synonyms: 4-Fluoro-2-methylbenzene-1-sulphonyl chloride

558567-1G 558567-5G
220078-408EA 32.88 USD
220078-408 220078-409
4-Fluoro-2-methylbenzenesulfonyl chloride ≥97%
4-Fluoro-2-methylbenzenesulphonyl chloride
Formula: C₇H₆ClFO₂S
MW: 208.64
Boiling Pt: 245…246 °C (1013 hPa)
Density: 1.433
MDL Number: MFCD03094390
CAS Number: 7079-48-3
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR