3-Fluoro-4-methylbenzenesulfonyl chloride ≥97%

Synonyms: 3-Fluoro-4-methylbenzene-1-sulphonyl chloride

558559-1G 558559-5G
220078-406EA 229.62 USD
220078-406 220078-407
3-Fluoro-4-methylbenzenesulfonyl chloride ≥97%
3-Fluoro-4-methylbenzenesulphonyl chloride
Formula: C₇H₆ClFO₂S
MW: 208.64 g/mol
Melting Pt: 46…50 °C
MDL Number: MFCD03094389
CAS Number: 90260-13-2
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR