2-Bromo-5-(trifluoromethyl)benzenesulfonyl chloride ≥97%

Synonyms: 2-Bromo-5-(trifluoromethyl)benzene-1-sulphonyl chloride

558435-1G 558435-5G
220078-396EA 34.46 USD
220078-396 220078-397
2-Bromo-5-(trifluoromethyl)benzenesulfonyl chloride ≥97%
2-Bromo-5-(trifluoromethyl)benzenesulphonyl chloride
Formula: C₇H₃BrClF₃O₂S
MW: 323.52 g/mol
Boiling Pt: 230…231 °C (1013 hPa)
MDL Number: MFCD03094395
CAS Number: 176225-08-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR