1-Boc-5-bromoindole

Synonyms: N-Boc-5-bromoindole, 5-Bromo-1-tert-butoxycarbonylindole, tert-Butyl 5-bromo-1H-indole-1-carboxylate

557749-25G-A 557749-5G-A
220078-349EA 602 USD
220078-349 220078-350
1-Boc-5-bromoindole
1-Boc-5-bromoindole
Formula: C₁₃H₁₄BrNO₂
MW: 296.16 g/mol
MDL Number: MFCD02090067
CAS Number: 182344-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR