2-Chloro-4-nitropyridine

557390-5G 557390-1G
220077-906EA 56.97 USD
220077-906 220077-905
2-Chloro-4-nitropyridine
2-Chloro-4-nitropyridine
Formula: C₅H₃ClN₂O₂
MW: 158.54 g/mol
Melting Pt: 53.5 - 54 °C
MDL Number: MFCD00661454
CAS Number: 23056-36-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR