4-(Bromomethyl)benzenesulfonyl chloride ≥95%

Synonyms: 4-(Bromomethyl)benzene-1-sulphonyl chloride

557153-1G 557153-5G
220078-333EA 37.48 USD
220078-333 220077-883
4-(Bromomethyl)benzenesulfonyl chloride ≥95%
4-(Bromomethyl)benzenesulphonyl chloride
Formula: C₇H₆BrClO₂S
MW: 269.55 g/mol
Melting Pt: 74 - 76 °C
MDL Number: MFCD00156129
CAS Number: 66176-39-4
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR