4-Bromophenyl methyl sulfone ≥97%

Synonyms: 1-Bromo-4-(methylsulphonyl)benzene

556327-5G 556327-25G
220076-934EA 80.88 USD
220076-934 220076-933
4-Bromophenyl methyl sulfone ≥97%
4-Bromophenyl methyl sulphone
Formula: C₇H₇BrO₂S
MW: 235.1 g/mol
MDL Number: MFCD00025065
CAS Number: 3466-32-8
EINECS: 222-421-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR