α-Bromo-2-chloro-5-fluorotoluene ≥99%

556319-10G 556319-1G
220076-931EA 295.08 USD
220076-931 220076-932
α-Bromo-2-chloro-5-fluorotoluene ≥99%
α-Bromo-2-chloro-5-fluorotoluene
Formula: C₇H₅BrClF
MW: 223.47 g/mol
MDL Number: MFCD01631419
CAS Number: 81778-09-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR