3-Methoxybenzenesulfonyl chloride ≥96%

Synonyms: 3-Methoxybenzene-1-sulphonyl chloride

556165-5G 556165-1G
220078-227EA 199.23 USD
220078-227 220078-226
3-Methoxybenzenesulfonyl chloride ≥96%
3-Methoxybenzenesulphonyl chloride
Formula: C₇H₇ClO₃S
MW: 206.65 g/mol
Boiling Pt: 86…98 °C (1 mmHg)
Density: 1.46 g/cm³ (20 °C)
MDL Number: MFCD01318155
CAS Number: 10130-74-2
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR