2-Cyanobenzenesulfonyl chloride ≥98%

Synonyms: 2-Cyanobenzene-1-sulphonyl chloride

556157-1G 556157-5G
220078-224EA 36.62 USD
220078-224 220078-225
2-Cyanobenzenesulfonyl chloride ≥98%
2-Cyanobenzenesulphonyl chloride
Formula: C₇H₄ClNO₂S
MW: 201.63 g/mol
Melting Pt: 67.5 °C
MDL Number: MFCD00052341
CAS Number: 69360-26-5
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR