4-Bromo-3-methylbenzenesulfonyl chloride ≥97%

Synonyms: 4-Bromo-3-methylbenzene-1-sulphonyl chloride

556009-5G 556009-1G
220078-134EA 318.77 USD
220078-134 220078-133
4-Bromo-3-methylbenzenesulfonyl chloride ≥97%
4-Bromo-3-methylbenzenesulphonyl chloride
Formula: C₇H₆BrClO₂S
MW: 269.55 g/mol
Melting Pt: 61 °C
MDL Number: MFCD03094696
CAS Number: 72256-93-0
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR