3,5-Difluorobenzenesulfonyl chloride ≥97%

Synonyms: 3,5-Difluorobenzene-1-sulphonyl chloride

555975-5G 555975-1G
220078-129EA 47.49 USD
220078-129 220078-128
3,5-Difluorobenzenesulfonyl chloride ≥97%
3,5-Difluorobenzenesulphonyl chloride
Formula: C₆H₃ClF₂O₂S
MW: 212.6 g/mol
MDL Number: MFCD02091378
CAS Number: 210532-25-5
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR