2-Bromo-4,6-difluorobenzenesulfonyl chloride ≥97%

555924-5G 555924-1G
220078-123EA 188.46 USD
220078-123 220078-122
2-Bromo-4,6-difluorobenzenesulfonyl chloride ≥97%
2-Bromo-4,6-difluorobenzenesulphonyl chloride
Formula: C₆H₂BrClF₂O₂S
MW: 291.5 g/mol
Boiling Pt: 265 - 267 °C (1013 hPa)
Density: 1.93 g/cm³ (20 °C)
MDL Number: MFCD03094561
CAS Number: 351003-42-4
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR