3,4-Difluorobenzenesulfonyl chloride ≥97%

Synonyms: 3,4-Difluorobenzene-1-sulphonyl chloride

555916-1G 555916-5G
220078-118EA 56.75 USD
220078-118 220078-119
3,4-Difluorobenzenesulfonyl chloride ≥97%
3,4-Difluorobenzenesulphonyl chloride
Formula: C₆H₃ClF₂O₂S
MW: 212.6 g/mol
Boiling Pt: 105 °C (15 mmHg)
Density: 1.586 g/cm³ (20 °C)
MDL Number: MFCD00153120
CAS Number: 145758-05-0
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR