2,4,6-Trichlorobenzenesulfonyl chloride ≥96%

Synonyms: 2,4,6-Trichlorobenzene-1-sulphonyl chloride

555649-1G 555649-5G
220078-108EA 37.91 USD
220078-108 220078-109
2,4,6-Trichlorobenzenesulfonyl chloride ≥96%
2,4,6-Trichlorobenzenesulphonyl chloride
Formula: C₆H₂Cl₄O₂S
MW: 279.96 g/mol
Melting Pt: 49 °C
MDL Number: MFCD00052973
CAS Number: 51527-73-2
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR