4-((Trifluoromethyl)thio)benzoyl chloride ≥97%

555479-5G 555479-1G
220077-881EA 60.42 USD
220077-881 220077-880
4-((Trifluoromethyl)thio)benzoyl chloride ≥97%
4-((Trifluoromethyl)thio)benzoyl chloride
Formula: C₈H₄ClF₃OS
MW: 240.63 g/mol
Boiling Pt: 96 °C (18 mmHg)
Density: 1.43 g/cm³ (20 °C)
MDL Number: MFCD01631632
CAS Number: 330-14-3
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR