4-Hydroxy-2-methoxybenzaldehyde

Warning

55543-5G 55543-1G
220077-876EA 117.38 USD
220077-876 220077-875
4-Hydroxy-2-methoxybenzaldehyde
4-Hydroxy-2-methoxybenzaldehyde
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 273 °C (1013 hPa)
Melting Pt: 156 - 158 °C
Storage Temperature: Ambient
MDL Number: MFCD00051964
CAS Number: 18278-34-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR