2,5-Difluorobenzenesulfonyl chloride ≥97%

Synonyms: 2,5-Difluorobenzene-1-sulphonyl chloride

555428-1G 555428-5G
220077-873EA 25.85 USD
220077-873 220077-874
2,5-Difluorobenzenesulfonyl chloride ≥97%
2,5-Difluorobenzenesulphonyl chloride
Formula: C₆H₃ClF₂O₂S
MW: 212.6 g/mol
MDL Number: MFCD00060682
CAS Number: 26120-86-5
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR