(4-Benzyloxyphenyl)acetic acid ≥98%

Synonyms: 2-(4-(Benzyloxy)phenyl)acetic acid

555398-5G 555398-1G
220077-868EA 118.85 USD
220077-868 220077-867
(4-Benzyloxyphenyl)acetic acid ≥98%
(4-Benzyloxyphenyl)acetic acid
Formula: (C₆H₅CH₂OC₆H₄)CH₂COOH
MW: 242.27 g/mol
MDL Number: MFCD00017540
CAS Number: 6547-53-1
EINECS: 229-463-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR