Cesium carbonate ≥99.95%

Warning

554855-50G 554855-10G
220078-064EA 466.15 USD
220078-064 220078-063
Cesium carbonate ≥99.95%
Cesium carbonate
Formula: Cs₂CO₃
MW: 325.82 g/mol
Melting Pt: 610 °C
Density: 4.24 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010957
CAS Number: 534-17-8
EINECS: 208-591-9
Merck Index: 13,02020

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR