(R)-(-)-2-Hydroxyisovaleric acid ≥98.0% (by titrimetric analysis)

Synonyms: D-α-Hydroxyisovaleric acid, 2-Hydroxyisovaleric-acid, (R)-2-Hydroxy-3-methylbutanoic acid, D-Valic acid, (2R)-2-Hydroxy-3-methylbutanoic acid

55452-1G 55452-5G
220078-037EA 238 USD
220078-037 220078-038
(R)-(-)-2-Hydroxyisovaleric acid ≥98.0% (by titrimetric analysis)
(R)-(-)-2-Hydroxyisovaleric acid
Formula: C₅H₁₀O₃
MW: 118.13 g/mol
MDL Number: MFCD00066442
CAS Number: 17407-56-6

Order Now

Specification Test Results

Assay 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR