Rubrene

Synonyms: 5,6,11,12-Tetraphenylnaphthacene, 5,6,11,12-Tetraphenyltetracene

554073-500MG 554073-100MG
220078-012EA 190.92 USD
220078-012 220078-011
Rubrene
Rubrene
Formula: C₄₂H₂₈
MW: 532.68 g/mol
MDL Number: MFCD00003703
CAS Number: 517-51-1
EINECS: 208-242-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR