4,5-Dicyanoimidazole

Synonyms: 4,5-Dicyano-1H-imidazole, Imidazole-4,5-dicarbonitrile

554030-5G 554030-25G 554030-1KG
220078-005EA 118.46 USD
220078-005 220078-004 220078-003
4,5-Dicyanoimidazole
4,5-Dicyanoimidazole
Formula: C₅H₂N₄
MW: 118.1 g/mol
Melting Pt: 176 °C
Density: 1.458 g/cm³ (20 °C)
MDL Number: MFCD00005194
CAS Number: 1122-28-7
EINECS: 214-344-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR