Magnesium diboride ≥96%

553913-5G 553913-25G
220078-000EA 95.38 USD
220078-000 220077-999
Magnesium diboride ≥96%
Magnesium diboride
Formula: B₂Mg
Density: 2.3 g/cm³ (20 °C)
MDL Number: MFCD00049752
CAS Number: 12007-25-9
EINECS: 234-501-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR