2,4-Dichloro-5-fluoro-3-nitrobenzoic acid ≥97%

553867-25G 553867-5G
220077-994EA 43.62 USD
220077-994 220077-995
2,4-Dichloro-5-fluoro-3-nitrobenzoic acid ≥97%
2,4-Dichloro-5-fluoro-3-nitrobenzoic acid
Formula: C₇H₂Cl₂FNO₄
MW: 254 g/mol
MDL Number: MFCD03427137
CAS Number: 106809-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR