3-Bromo-2-chloropyridine

Warning

553751-25G 553751-5G
220077-988EA 85.51 USD
220077-988 220077-989
3-Bromo-2-chloropyridine
3-Bromo-2-chloropyridine
Formula: C₅H₃BrClN
MW: 192.44 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 55 °C
MDL Number: MFCD00234007
CAS Number: 52200-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR